Learn More
Fisherbrand™ Polyolefin Cryogenic Label Sheets
Description
- Temperature resistance -196°C to 150°C.
- Liquid and vapor phase LN2, water baths to 100°C and autoclaving and moderate ovens to 150°C
- Chemically inert resistant to most solvents and caustic agents.
- Adhere to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling, or degrading
- Label sheets will not jam printers
- Flexible polyolefin material and acrylic adhesive
- Unique combo expands and contracts together with temperature fluctuations
- 20 sheets per pack
- Autoclavable
- Disposable
- Rainbow packs contain 4 sheets of each Blue, Green, Orange, Red, Yellow
Specifications
Specifications
| Finish | Matte |
| Format | Sheet |
| Material | Polyolefin, Acrylic |
| Print Technology | Laser |
| Product Type | Cryogenic Label |
| Sterility | Non-sterile |
| For Use With (Equipment) | Cryogenic vials |
| Labels per Sheet | 52 |
| No. per Pack | 1040 Labels |
| Shape | Rectangle |
| Show More |