missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B2 Formate Salt, TRC

Product Code. 30413166
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413166 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413166 Supplier Toronto Research Chemicals Supplier No. TRCP6893962.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polymyxin B2 Formate Salt
Purity Grade Notes HPLC
Molecular Formula C55H96N16O13 • x(HCOOH)
Quantity 2.5 mg
Synonym 1-[(2S)-4-Amino-2-[(6-methyl-1-oxoheptyl)amino]butanoic acid]polymyxin B1 Formate Salt, N2-(6-Methyl-1-oxoheptyl)-L-2,4-diaminobutanoyl-L-threonyl-L-2,4-diaminobutanoyl-L-2,4-diaminobutanoyl-L-2,4-diaminobutanoyl-D-phenylalanyl-L-leucyl-L-2,4-diaminobutanoyl-L-2,4-diaminobutanoyl-L-threonine (10-4)-lactam Formate Salt
SMILES O=C([C@H](CCN)NC1=O)N[C@H](CC2=CC=CC=C2)C(N[C@@H](CC(C)C)C(N[C@@H](CCN)C(N[C@@H](CCN)C(N[C@]([C@@H](C)O)([H])C(NCC[C@@H]1NC([C@H](CCN)NC([C@@]([C@@H](C)O)([H])NC([C@H](CCN)NC(CCCCCCC)=O)=O)=O)=O)=O)=O)=O)=O)=O.[HCOOH]
Molecular Weight (g/mol) 1189.45
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.