Learn More
Kinematica™ Polymix™ PX-MFC 90 D Hammer or Cutting Mill
Product Code. 11900359 Shop All Kinematica Products
Change view
Click to view available options
Product Type:
Blade Grinding Mill
Hammer Grinding Mill
Plug Type:
EU plug
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Plug Type Unit Size
11900359 Blade Grinding Mill EU plug Each
11910359 Hammer Grinding Mill EU plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11900359 Supplier Kinematica™ Supplier No. 35010031
  • ONLINE EXCLUSIVE PRICE
    8925.00 EUR
    8288.00 EUR / Pack of 1
    Save 637.00 EUR
    7% Off
View all promotions
Estimated Shipment: 13-11-2026
Polymix™ MFC 90 D features easy to change grinding attachments and sieves extending the range of samples that can be processed. Simple handling, high user safety and efficient grinding results are just a few of the advantages of this mill.
  • Working volume approx. 300mL (volume of the funnel)
  • 600W high frequency 3 phase motor with speed regulation from 50rpm up to 6, 000rpm
  • Integrated overheating, overload and blocking protection
  • Safety switch for automatic off
  • Energy saving through automatic standby mode
  • Hammer grinding or blade grinding attachment
  • Sieve size 2mm.
Specifications
Product Type Blade Grinding Mill
Certifications/Compliance IEC/EN 61000-6-2 / EN 61000-6-3, IEC/EN 61010-2-51
Dimensions (L x W x H) 325 x 251 x 480 mm
Electrical Requirements 230 V/EU, 50 Hz, 1'000 W Motor Input Power
Height (Metric) 480 mm
Hertz 50/60 Hz
IP Rating IP20
Length (Metric) 325 mm
Plug Type EU plug
Voltage 230 V
Watts 1000 W
Weight (Metric) 12 kg
Width (Metric) 251 mm