Learn More
Scilabub™ Polyimide Cryogenic Glove
Product Code. 12981185
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12981185 Large 1 pair
12342239 Medium 1 pair
12961185 Small 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12981185 Supplier Scilabub™ Supplier No. GLO/CMEL
  • ONLINE EXCLUSIVE PRICE
    270.00 EUR
    224.28 EUR / 1 pairs
    Save 45.72 EUR
    16% Off
Estimated Shipment: 22-12-2026
Polyimide Cryogenic Gloves have been specially developed for the handling of very cold objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
  • Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C)
  • Gloves should not actually be immersed in liquid nitrogen
  • The outer layer is waterproof material to resist condensed water on cold surfaces
  • Permeable to gases (including water vapour) for enhanced user comfort so that there is no clammy condensation inside
  • Stitch holes unsealed to reduce bulk around the fingers
  • Very durable, and retain flexibility (albeit with some stiffening) for long periods in cold atmospheres
  • Polyimide outer material, polyolefin with some polyester as filling and cotton liner
  • Manufactured to EN511
Specifications
Classification PPE Category III
Coating Material Polyimide
Color Blue
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gloves
Size Large
Sterility Non-sterile
Temperature (Metric) Max -190°C
Liner Material Cotton
Quantity 1 Pack
Resistant To Water
Length (Metric) 500 mm