Learn More
Scilabub™ Polyimide Cryogenic Glove
Product Code. 12961185
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12961185 Small 1 pair
12981185 Large 1 pair
12342239 Medium 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12961185 Supplier Scilabub™ Supplier No. GLO/CMES
  • ONLINE EXCLUSIVE PRICE
    270.00 EUR
    224.28 EUR / 1 pairs
    Save 45.72 EUR
    16% Off
Estimated Shipment: 07-01-2027
Polyimide Cryogenic Gloves have been specially developed for the handling of very cold objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
The outer layer is of waterproof material to resist condensed water on cold surfaces, but remains permeable to gases (including water vapour) so that there is no clammy condensation inside. To reduce bulk around the fingers, the stitch holes have not been sealed. In normal use this has not proved a problem, however, the gloves should not be immersed in liquid nitrogen and it is not advisable to use them above -80°C. Very durable, they retain their flexibility (albeit with some stiffening) for long periods in cold atmospheres. Polyimide outer material, polyolefin with some polyester as filling and cotton liner. Manufactured to EN511.Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C).
Specifications
Classification PPE Category III
Coating Material Polymide
Product Type Cryogenic Gloves
Size Small
Temperature (Metric) Max -190°C
Liner Material Cotton
Quantity 1 Pack
Resistant To Water
Length (Metric) 500 mm