Learn More
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials
Product Code. 15354789 Shop All Fisherbrand Products
Change view
Click to view available options
Closure Size:
8 mm
10 mm
22 mm
Unit Size:
Pack of 300
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
15354789 22 mm Pack of 300
15364789 8 mm Pack of 300
15374789 10 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15354789
  • ONLINE EXCLUSIVE PRICE
    31.95 EUR
    22.01 EUR / Pack of 300
    Save 9.94 EUR
    31% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Fit securely into place to seal glass specimen vials. Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials are available in a variety of sizes specifically designed to work with Fisherbrand™ Push-in Cap Glass Specimen Vials.
  • Specifically designed for use with Fisherbrand™ Push-in Cap Glass Specimen Vials
  • Polyethylene construction, natural color
  • Available in a variety of sizes to match vial specifications
  • Push-in cap specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Polyethylene
Closure Type Push-in Cap
Closure Size 22 mm