Learn More
Kartell™ Polyethylene Integral Eye Wash Bottle
Product Code. 10021071
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10021071 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10021071 Supplier Kartell™ Supplier No. 0038300
  • ONLINE EXCLUSIVE PRICE
    128.00 EUR
    126.15 EUR / Pack of 10
    Save 1.85 EUR
    1% Off
Estimated Shipment: 11-02-2027
Equipped with an eyecup and specially designed snap-on cap for fast and convenient eye washing to remove foreign objects quickly and prevent serious damage to vision. Kartell™ Polyethylene Integral Eye Wash Bottles can be filled with distilled water or any other medicated liquid intended for ophthalmic use.
Equipped with an eyecup and specially designed snap-on cap for fast and convenient eye washing to remove foreign objects quickly and prevent serious damage to vision. Kartell™ Polyethylene Integral Eye Wash Bottles can be filled with distilled water or any other medicated liquid intended for ophthalmic use.
  • Automatic valve incorporated in the head to admmit air and prevent contaminated eyewash from flowing back into bottle
  • Perforated plate in eyecup breaks up the stream and provides a uniform spray to the eye
  • Includes a tube to drain the contaminated eyewash
  • Suitable for foodstuff
Specifications
Capacity (Metric) 500 mL
Height (Metric) 213 mm
Material Polyethylene
Product Type Integral Eye Wash Bottle
Width (Metric) 73 mm