Learn More
Cole-Parmer™ Glove Bag™ Inflatable Glove Chambers
Product Code. 12318627
Change view
Click to view available options
Length (Metric):
431.8 mm
508 mm
685.8 mm
939.8 mm
Width (Metric):
431.8 mm
508 mm
635 mm
685.8 mm
939.8 mm
Height (Metric):
279.4 mm
304.8 mm
381 mm
457.2 mm
939.8 mm
Unit Size:
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Height (Metric) unitSize
12318627 431.8 mm 431.8 mm 279.4 mm Pack of 6
12328627 431.8 mm 685.8 mm 304.8 mm Pack of 6
12368627 508 mm 508 mm 304.8 mm Pack of 6
12338627 685.8 mm 685.8 mm 381 mm Pack of 6
12358627 939.8 mm 635 mm 939.8 mm Pack of 6
12348627 685.8 mm 939.8 mm 457.2 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12318627 Supplier Cole-Parmer™ Supplier No. 0440830
  • ONLINE EXCLUSIVE PRICE
    448.73 EUR
    357.24 EUR / Pack of 6
    Save 91.49 EUR
    20% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Protect yourself and your materials quickly without giving up lab space with these glove chambers. Glove Bag™ Inflatable Glove Chambers establish high-purity working atmosphere in minutes. Inflatable chambers effectively eliminate oxygen and moisture during material manipulation operation.
An alternative to standard glove boxes—these durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeves, seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate.
  • Disposable chambers simplify cleanup
  • Two equipment sleeves and two rear-mounted gas ports for added versatility
  • Glove bags have a 2.5 mil thickness and are made of polyethylene
  • Glove bags are not autoclaveable, nor are they sterile
  • One pair of cotton gloves, tubing coupler and a closure included
This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
Material Polyethylene
Height (Metric) 279.4 mm
Includes One Pair Of Cotton Gloves, Tubing Coupler, and a Closure
Length (Metric) 431.8 mm
Quantity 6/Pk.
Width (Metric) 431.8 mm