Learn More
Sperian™ Polyester Webbing Anchorage
Product Code. 12652793
Change view
Click to view available options
Length (Metric):
1.2 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12652793 1.2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12652793 Supplier Sperian™ Supplier No. 1002918
  • ONLINE EXCLUSIVE PRICE
    34.65 EUR
    15.04 EUR / Pack of 1
    Save 19.61 EUR
    56% Off
Estimated Shipment: 23-03-2027
Specifications
Certifications/Compliance ISO 9001 / 2000
For Use With (Application) The versatile hurokheveder be used for many mooring points.
Weight (Metric) 0.21kg
Length (Metric) 1.2 m
Material polyester
Product Type Webbing Anchorage