Learn More
Brady™ Polyester Labels
Product Code. 13521460 Shop All Brady Products
Change view
Click to view available options
Color:
Transparent
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13521460 White Each
13531460 Transparent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13521460 Supplier Brady™ Supplier No. 143081
  • ONLINE EXCLUSIVE PRICE
    173.00 EUR
    155.47 EUR / Pack of 1
    Save 17.53 EUR
    10% Off
In Stock
Polyester Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant.
Specifications
Certifications/Compliance Halogen Free (DIN VDE 0472 Part 815)
Description Polyester Labels for BBP33/i3300 Printers
Adhesive Type Acrylic, Permanent
Coating None
Color White
Display Blank
Finish Gloss
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Barcode labelling, Circuit board labelling, Component and equipment labelling, General Identification, Inventory and inspection labelling
For Use With (Equipment) i3300, i5300, BBP33
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
No. per Pack 1500 Labels/Box
Quantity 1500 Pk.
Shape Rectangle
Size (Metric) 76.20 x 25.4 mm (W x H)
Software Type LabelMark, MarkWare
Temperature Range (English) -94°F to +230°F
Temperature Range (Metric) -70°C to +110°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.076 mm
Vertical Repeat (Metric) 28.58 mm
Dimensions (L x W) 25.4 x 76.2 mm
Length (English) 1 in.
Length (Metric) 25.4 mm
Width (English) 3 in.
Width (Metric) 76.2 mm
Height (Metric) 25.40 mm
Alternate Part Number B33-18-423