Learn More
Brady™ Polyester High Performance Tape
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent, Pressure-sensitive |
| Color | Tan |
| Material | Polyester |
| Product Type | High Performance Labelling Tape |
| For Use With (Application) | For use on Stainless Steel pipes in harsh environments |
| Gloss | 80 Gardner Units |
| Quantity | 750/Pk. |
| Thickness (Metric) | 0.019 mm |
| Dimensions (L x W) | 30 m x 13 mm |
| Length (Metric) | 30 m |
| Show More |