missing translation for 'onlineSavingsMsg'
Learn More

Polybutylene Terephthalate Cyclic Trimer (Cyclotris(1,4-butylene Terephthalate)), Mikromol™

Product Code. 17937304
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937304 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17937304 Supplier Mikromol Supplier No. MM3898.020025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Polybutylene Terephthalate Cyclic Trimer (Cyclotris(1,4-butylene Terephthalate))
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclotris(1,4-butylene Terephthalate)
CAS 63440-94-8
Analyte or Component Names Cyclotris(1,4-butylene Terephthalate)
Shipping Condition Room Temperature
Molecular Formula C36 H36 O12
Certifications/Compliance ISO 9001
Synonym 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-10,12,22,24,34,36,37,39,41-nonaene-2,9,14,21,26,33-hexone, PBT Cyclic Trimer, 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-1(36),10,11,13(12),22,23,25(24),34,37-nonaene-2,9,14,21,26,33-hexone
InChI Formula InChI=1S/C36H36O12/c37-31-25-7-9-27(10-8-25)33(39)45-21-3-4-23-47-35(41)29-15-17-30(18-16-29)36(42)48-24-6-5-22-46-34(40)28-13-11-26(12-14-28)32(38)44-20-2-1-19-43-31/h7-18H,1-6,19-24H2
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc(cc3)C(=O)OCCCCOC(=O)c4ccc1cc4
Molecular Weight (g/mol) 660.66
Quantity 25 mg
Formula Weight 660.2207 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.