missing translation for 'onlineSavingsMsg'
Learn More

Polybutylene Terephthalate Cyclic Tetramer (Cyclotetrakis(1,4-butylene Terephthalate)), Mikromol™

Product Code. 17959884
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959884 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17959884 Supplier Mikromol Supplier No. MM3898.030025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Polybutylene Terephthalate Cyclic Tetramer (Cyclotetrakis(1,4-butylene Terephthalate))
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polybutylene Terephthalate Cyclic Tetramer
CAS 29278-72-6
Analyte or Component Names Polybutylene Terephthalate Cyclic Tetramer
Shipping Condition Room Temperature
Molecular Formula C48 H48 O16
Certifications/Compliance ISO 9001
Synonym 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,11,12,22,23,24,34,35,36,46,48,49-dodecaene-2,9,14,21,26,33,38,45-octone, 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,12,22,24,34,36,46,48,49,51,53,55-dodecaene-2,9,14,21,26,33,38,45-octone (9CI), Terephthalic acid, tetramol. cyclic tetramethylene ester (8CI), 1,4-Butanediol, tetramol. cyclic terephthalate (8CI), Cyclotetrakis(1,4-butylene Terephthalate), PBT Cyclic Tetramer, Polybutylene Terephthalate Cyclic Tetramer (Cyclotetrakis(1,4-butylene Terephthalate))
InChI Formula InChI=1S/C48H48O16/c49-41-33-9-11-35(12-10-33)43(51)59-27-3-4-29-61-45(53)37-17-19-39(20-18-37)47(55)63-31-7-8-32-64-48(56)40-23-21-38(22-24-40)46(54)62-30-6-5-28-60-44(52)36-15-13-34(14-16-36)42(50)58-26-2-1-25-57-41/h9-24H,1-8,25-32H2
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc(cc3)C(=O)OCCCCOC(=O)c4ccc(cc4)C(=O)OCCCCOC(=O)c5ccc1cc5
Molecular Weight (g/mol) 880.8851
Quantity 25 mg
Formula Weight 880.2942 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.