Learn More
Sperian™ Polyamide Anchorage Sling
Product Code. 12684983
Change view
Click to view available options
Length (Metric):
0.6 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12684983 0.6 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12684983 Supplier Sperian™ Supplier No. 1008360
  • ONLINE EXCLUSIVE PRICE
    17.85 EUR
    8.48 EUR / Pack of 1
    Save 9.37 EUR
    52% Off
Estimated Shipment: 08-02-2027
Specifications
Certifications/Compliance ISO 9001 / 2000
Dimensions (L x W) 0.6 m x 25 mm
For Use With (Application) Titan webbing anchorage sling adapts to a wide range of anchorage points.
Weight (Metric) 0.08kg
Width (Metric) 25 mm
Length (Metric) 0.6 m
Material Polyamide
Product Type Anchorage Sling