Learn More
Koflo Unmounted In-Line Static Mixers
Product Code. 11792653 Shop All Cole Parmer Products
Change view
Click to view available options
Length (Metric):
121 mm
127 mm
152 mm
241 mm
254 mm
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11792653 127 mm Pack of 10
11754109 121 mm Pack of 10
11744109 152 mm Pack of 10
11764109 241 mm Pack of 10
11774109 254 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11792653 Supplier Koflo Supplier No. 0466708
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Unmounted In-Line Static Mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table.
  • They work well with flexible tubing.
Specifications
Diameter (English) 0.5 in.
Diameter (Metric) 13 mm
Length (English) 5 in.
Length (Metric) 127 mm
Type Unmounted In-line Mixer