missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene), TRC

Product Code. 30138120
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138120 500 g 500g
30136994 250 g 250g
30005195 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138120 Supplier Toronto Research Chemicals Supplier No. TRCP689465500G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-53-6
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
Synonym Polystyrene
IUPAC Name styrene
SMILES C=CC1=CC=CC=C1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2
Formula Weight 104.063
Chemical Name or Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.