Learn More
MP Biomedicals™ Poly-L-Glutamic Acid, MW 50,000-100,000
Product Code. 11447162 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1000 mg
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447162 1000 mg 1g
11437162 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11447162 Supplier MP Biomedicals™ Supplier No. 0215191891
  • ONLINE EXCLUSIVE PRICE
    1360.00 EUR
    1294.65 EUR / 1g
    Save 65.35 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Specifications
Purity 95%
Content And Storage 0°C
Format Powder
For Use With (Application) Used in the construction of Polypeptide based Copolymer membranes, micelles and nanoparticles
Molecular Weight (g/mol) 50000 to 100000
Product Type Poly-L-Glutamic Acid from Synthetic
Quantity 1000 mg