Learn More
Cytiva Poly(I)-Poly(C)
Product Code. 11545784 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
265 mg
Unit Size:
265mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11545784 265 mg 265mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11545784 Supplier Cytiva Supplier No. 27473201
  • ONLINE EXCLUSIVE PRICE
    2780.60 EUR
    2350.48 EUR / 265mg
    Save 430.12 EUR
    15% Off
 Restricted Item
In Stock
For stimulation of interferon production in vitro. Cytiva Poly(I)-Poly(C) is supplied as a sterile lyophilised powder.
  • Double-stranded polyribonucleotide poly(I)−poly(C) has been shown to induce interferon in rabbit cells
  • This product is dispensed by weight of dry material
  • This weight includes polymer, residual salt and residual water
  • Content of polymer may vary from lot to lot
TRUSTED_SUSTAINABILITY
Specifications
Quantity 265 mg
Color White
Form Powder