missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
POLR1D Polyclonal antibody specifically detects POLR1D in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | POLR1D |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH7.0 |
| Gene Alias | AC19, DNA-directed RNA polymerase I subunit D, DNA-directed RNA polymerases I and III subunit RPAC2, FLJ20616, hRPA19, MGC9850, POLR1C, polymerase (RNA) I polypeptide D, 16kDa, RPA16RNA polymerase I 16 kDa subunit, RPA9, RPAC2RNA polymerases I and III subunit AC2, RPC16, RPO1-3, TCS2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human POLR1D fragment |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?