Learn More
Poloxamer Solid, TRC
Product Code. 30138165
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138165 100 g 100g
30012161 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138165 Supplier Toronto Research Chemicals Supplier No. TRCP688040100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9003-11-6
Molecular Formula ((C2 H4 O)mon(C3 H6 O)mon)ran
Molecular Weight (g/mol) 102.13
Synonym Propylene oxide, polymer with ethylene oxide
IUPAC Name 2-methyloxirane
SMILES [H]OCCOC(C)COCCO
Specifications
Quantity 100 g
Formula Weight 102.0681
Chemical Name or Material Poloxamer