missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 5.2, RNAse free

Product Code. 16825771
Change view
Click to view available options
Quantity:
100 mL
125 mL
250 mL
Unit Size:
100mL
125mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16825771 100 mL 100mL
15470217 125 mL 125mL
15480217 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16825771 Supplier Thermo Scientific Alfa Aesar Supplier No. J61928.AE
  • ONLINE EXCLUSIVE PRICE
    61.25 EUR
    54.65 EUR / 100mL
    Save 6.60 EUR
    10% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-09-3 ∣ C2H3NaO2 ∣ 82.03 g/mol

Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

Solubility

Soluble in water (246 g/L) at 20°C, and ethanol.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

pH 5.2
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Linear Formula NaOOCCH3
Solubility Information Soluble aq. soln. (246 g/L) (at 20°C,and ethanol.)
Formula Weight 82.03
Grade Ungraded
Concentration or Composition (by Analyte or Components) 3 M aq. soln.
Chemical Name or Material Sodium acetate, RNAse free

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.