Learn More
Thermo Scientific™ PolarScreen™ ER Beta Competitor Assay Kit, Red
Description
The PolarScreen™ ER Beta Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential estrogen receptor (ER) beta ligands. The kit uses insect cell-expressed, full-length, native (untagged) human ER beta (Cat. No. A15664) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone™ ES2) in a homogenous, mix-and-read assay format.
This kit produces a red color read-out; a 'green' kit is also available (Cat. No. A15890). Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance email drugdiscoverytech@lifetech.com.
The PolarScreen™ ER Beta Competitor Assay Kit is:
- Complete & ready-to-use out of the box—all reagents included, just add your test compounds
- High throughput compatible—20 μL fluorescence polarization assays in a 384-well plate
- Convenient—available in bulk sizes for larger screening needs
How the Assay Works
Full-length ER beta is added to a fluorescent estrogen ligand to form an ER-Fluormone™ ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone™ ES2 ligand from ER beta, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER beta.
All components are manufactured under the strictest quality parameters. This product's performance, per the Certificate of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
Specifications
Specifications
| Color | Red |
| Description | PolarScreen™ ER Beta Competitor Assay Kit, 1 Kit, Red, 384-Well Plate Format, Fluorescence Polarization Technique, Estrogen Receptor Beta Nuclear Receptor Target Gene, For Pharma & Biopharma, Target & Lead Identification & Validation |
| Includes | Estrogen receptor (ER) beta recombinant protein, full length, Fluromone™ EL red, ER red assay buffer |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Packaging | 384-well Plate |
| Content And Storage | • Estrogen Receptor (ER) Beta Recombinant Protein, Full Length (store at -68°C to -85°C) • Fluromone™ EL Red (-5°C to -30° C) • ER Red Assay Buffer (room temperature) |
| Assay Category | Biochemical Assay |
| Druggable Target | Nuclear Receptors |
| Readout | Fluorescence Polarization |
| Show More |