missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNU-120596, MedChemExpress
Description
PNU-120596 (NSC 216666) is a potent and selective α7 nAChR positive allosteric modulator (PMA) with an EC50 of 216 nM. PNU-120596 is inactive against α4β2, α3β4, and α9α10 nAChRs. PNU-120596 has the potential for psychiatric and neurological disorders research.
Specifications
Specifications
| Chemical Name or Material | PNU-120596 |
| CAS | 501925-31-1 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C13H14ClN3O4 |
| Quantity | 100 mg |
| Synonym | NSC 216666 |
| Solubility Information | DMSO : 50 mg/mL (160.40 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC(Cl)=C(C=C1OC)OC)NC2=NOC(C)=C2 |
| Molecular Weight (g/mol) | 311.72 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?