Description
- Cable ducts allow clean, safe installation of electrical cables in the hood workspace
- Provide chemical resistance and visual inspection of cables
- Avoid cable clutter and protect against mechanical wear and contamination
- Mounted on the right side of the hood for ergonomic access and optimal cable management
- Improve airflow fluidity by reducing obstruction caused by cables entering directly through the chassis
- Ideal for configurations involving multiple instruments, sensors or monitoring devices
Specifications
Specifications
| For Use With (Equipment) | Hoods and Enclosures |
| Product Type | Upper Duct Transition Adapter |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?