missing translation for 'onlineSavingsMsg'
Learn More

Plumbagin, TRC

Product Code. 30003558
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003558 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003558 Supplier Toronto Research Chemicals Supplier No. TRCP627000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 481-42-5
Molecular Formula C11 H8 O3
Quantity 100 mg
Synonym 1,4-Naphthalenedione, 5-hydroxy-2-methyl-, 1,4-Naphthoquinone, 5-hydroxy-2-methyl- (7CI,8CI), Plumbagin (6CI), 2-Methyl-5-hydroxy-1,4-naphthalenedione, 2-Methyl-5-hydroxy-1,4-naphthoquinone, 2-Methyljuglone, 5-Hydroxy-2-methyl-1,4-naphthalenedione, 5-Hydroxy-2-methyl-1,4-naphthoquinone, NSC 236613, NSC 688284, Plumbagine, Plumbagone
InChI Formula InChI=1S/C11H8O3/c1-6-5-9(13)10-7(11(6)14)3-2-4-8(10)12/h2-5,12H,1H3
SMILES CC1=CC(=O)c2c(O)cccc2C1=O
IUPAC Name 5-hydroxy-2-methylnaphthalene-1,4-dione
Molecular Weight (g/mol) 188.18
Formula Weight 188.0473
Chemical Name or Material Plumbagin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.