Learn More
Hamilton™ Plug 1/4-28 Threads 10/pk
Description
Specification Sheet
- Connection 1 – Male 1/4-28
- Connection 2 – Plug
- Fitting Material – CTFE
- Maximum Pressure – 87 psi (6 bar)
- Sales Quantity – 10 per pack
- I.D. – Plug
Specifications
Specifications
| Type | Thread |
| Quantity | 10/Pk. |
| Material | CTFE |