missing translation for 'onlineSavingsMsg'
Learn More

Plerixafor, TRC

Product Code. 17921625
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921625 100 mg 100mg
17901455 50 mg 50mg
17911525 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17921625 Supplier Toronto Research Chemicals Supplier No. TRCP580500100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110078-46-1
Molecular Formula C28 H54 N8
Molecular Weight (g/mol) 502.78
Synonym 1,4,8,11-Tetraazacyclotetradecane, 1,1'-[1,4-phenylenebis(methylene)]bis-, 1,1'-[1,4-Phenylenebis(methylene)]bis[1,4,8,11-tetraazacyclotetradecane], 1,1'-Xylyl bis-1.4,8,11 tetraaza cyclotetradecane, AMD 3100, JKL 169, JM 3100, Mozobil, Plerixafor, SDZ SID 791, 1,4,8,11-Tetraazacyclotetradecane, 1,1'-[1,4-phenylenebis(methylene)]bis-, 1,1'-[1,4-Phenylenebis(methylene)]bis[1,4,8,11-tetraazacyclotetradecane], 1,1'-Xylyl bis-1.4,8,11 tetraaza cyclotetradecane, AMD 3100, JKL 169, JM 3100, Mozobil, Plerixafor, SDZ SID 791, SID791
IUPAC Name 1-[[4-(1,4,8,11-tetrazacyclotetradec-1-ylmethyl)phenyl]methyl]-1,4,8,11-tetrazacyclotetradecane
SMILES C(N1CCCNCCNCCCNCC1)c2ccc(CN3CCCNCCNCCCNCC3)cc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C28H54N8/c1-9-29-15-17-31-13-3-21-35(23-19-33-11-1)25-27-5-7-28(8-6-27)26-36-22-4-14-32-18-16-30-10-2-12-34-20-24-36/h5-8,29-34H,1-4,9-26H2
Formula Weight 502.4471
Chemical Name or Material Plerixafor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.