Learn More
Fisherbrand™ Replacement Electrode Wire
Product Code. 11887803 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
1m x 0.2 mm
500 x 0.2 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11887803 1m x 0.2 mm Each
11817583 500 x 0.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11887803
  • ONLINE EXCLUSIVE PRICE
    260.00 EUR
    170.94 EUR / Pack of 1
    Save 89.06 EUR
    34% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
Replacement Electrode Wire is an accessoty for use with Verti-Gel Maxi, 2-Gel and 4-Gel Electrophoresis System.
  • Made of platinum
Specifications
For Use With (Equipment) Vertical Gel Maxi Electrophoresis Tank
Type Electrode Wire Replacement
Material Platinum
Thickness (Metric) 0.2 mm
Dimensions (L x W) 1m x 0.2 mm
Length (Metric) 100 cm