missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Platinum Iridium foil, 0.05mm (0.002 in.) thick, 99.9% (metals basis)
Specifications
Specifications
| Chemical Name or Material | Platinum Iridium foil |
| Name Note | 0.05mm (0.002 in.) thick |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 80:20 wt% |
| Quantity | 50 x 50 mm |
| Odor | Odorless |
| Weight | ≈0.68g/25x25mm |
| Form | Foil |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD02091683 |
| Percent Purity | 99.9% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?