Learn More
Platinum(II) Chloride (73% Pt), Sigma-Aldrich™
Specifications
Specifications
| Melting Point | 581°C (Decomposes) |
| CAS | 10025-65-7 |
| Quantity | 500 mg |
| Density | 6.1 g/cm3 |
| MDL Number | MFCD00011181 |
| Synonym | Platinum(II) chloride (73% Pt), Platinum dichloride, Platinum(II) chloride (73% Pt) |
| Molecular Weight (g/mol) | 265.98 |
| Grade | Research |
| Chemical Name or Material | Platinum(II) Chloride (73% Pt) |