Learn More
Gilson™ PLATEMASTERTM Height Stop
Product Code. 15823192 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
PLATEMASTER™
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15823192 PLATEMASTER™ 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15823192 Supplier Gilson™ Supplier No. F1077605
  • ONLINE EXCLUSIVE PRICE
    44.15 EUR
    40.95 EUR / 1 sets
    Save 3.20 EUR
    7% Off
In Stock
Easily adjust pipetting height with this pipetting head height adjuster.
The Height Stop is an accessory for the PLATEMASTERTM allowing for easy altering of pipetting height.
  • Ensures appropriate tip depth within shallow- or deep-well microplates
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipetter Accessory
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Color Black
Material Polypropylene
For Use With (Application) Liquid Handling
For Use With (Equipment) PLATEMASTER™
Quantity 1
Warranty 1 Year
Width (English) 2.76 in
Height (English) .51 in
Height (Metric) 13 mm
Weight (English) .07 lb
Weight (Metric) .03 kg
No. per Pack 1