Learn More
BD Plastipak™ Centric Luer-lock Hypodermic Three-Piece Syringe
Product Code. 11303040
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
Unit Size:
Pack of 125
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11303040 3 mL Pack of 200
11364881 5 mL Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11303040 Supplier BD Supplier No. 309658
  • ONLINE EXCLUSIVE PRICE
    123.80 EUR
    40.02 EUR / Pack of 200
    Save 83.78 EUR
    67% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Plastic graduated syringe. BD Medical™ Plastipak™ Centric Luer-lock Hypodermic Three-Piece Syringe for injection of fluid.
  • Plastic syringe
  • Includes luer-lock
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance NF EN 20594-1, ISO 594-1 de December 1993, ISO 594-2 de september 1998
Material Polypropylene
Diameter (Metric) Outer 10.12 mm
Latex Free Yes
Length (Metric) 69 mm
Product Type Centic Luer-lock Hypodermic 3-piece Syringe
Disposable Single-use
Graduations 0.1 mL
Quantity 200/Pk.
Volume (Metric) 3 mL
Connection Type Centric Luer Lock
Graduated Yes