Learn More
Scientific Industries SI™ Replacement Platform with Rubber Cover
Fits Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers. Scientific Industries SI™ Replacement Platform without Rubber Cover is for use with vortexing in beakers, flasks and multiple tubes.
Brand: Scientific Industries SI™ 0K-0500-902
Additional Details : Weight : 0.62000kg
Descripción
- For use with Vortex-Genie family of products
- Cover not included
Especificaciones
Molded rubber | |
Vortex-Genie family of mixers |