Learn More
Plas Labs™ Broome-Style Rodent Restrainers
Product Code. 15193745
Change view
Click to view available options
Diameter (English):
1 in.
1.5 in.
2 in.
2.5 in.
Length (English):
3.75 in.
5.5 in.
8 in.
8.5 in.
Weight Range:
125 to 250 g
15 to 30 g
250 to 500 g
30 to 125 g
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Length (English) Weight Range Unit Size
15193745 2.5 in. 8.5 in. 250 to 500 g Each
12505846 1 in. 3.75 in. 15 to 30 g Each
12824593 1.5 in. 5.5 in. 30 to 125 g Each
13204979 2 in. 8 in. 125 to 250 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15193745 Supplier Plas Labs™ Supplier No. 554BSRR
  • ONLINE EXCLUSIVE PRICE
    226.80 EUR
    138.17 EUR / Pack of 1
    Save 88.63 EUR
    39% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Ideal for injections or blood sampling
  • Offer easy access to rodent
  • Prevent rodent from turning
  • Provide safety to the animal handler
  • Manufactured from clear materials to provide complete visibility of animal
Specifications
Color Clear
For Use With (Application) Injections or Blood Sampling
Type Rodent Restrainer
Material Noryl™ Plastic
Sterilization Method Chemically sterilized or washed at 180°F
Style Broome
Length (English) 8.5 in.
Length (Metric) 21.59 cm
Diameter (English) 2.5 in.
Diameter (Metric) 6.35 cm
Weight Range 250 to 500 g