missing translation for 'onlineSavingsMsg'
Learn More

Placental Lactogen (hPL) Rabbit Polyclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.

109.14 EUR - 374.00 EUR

Specifications

Antigen Placental Lactogen (hPL)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 4
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12633437
View Documents
Epredia
RB-9067-P0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12643437
View Documents
Epredia
RB-9067-P1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
229.00 EUR
208.83 EUR / 0.50mL
Save 20.17 EUR
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12623437
View Documents
Epredia
RB-9067-P
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12673437
View Documents
Epredia
RB-9067-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Human placental lactogen (hPL) is known to originate in the syncytiotrophoblasts. Ultrastructural localization indicates that placental lactogen is stored in small morphologically distinct cytoplasmic granules within these cells and has a different secretory mechanism from other hormonal peptides such as human chorionic gonadotropin. The level of expression of placental lactogen through pregnancy increases gradually to term.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human placental lactogen

Molecular Weight: 21kDa

Positive Control: hPL recombinant protein, Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Placental Lactogen (hPL)
Polyclonal
Rabbit
Steroids Hormones and Growth Factors
Purified human placental lactogen
Primary
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IVD
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.