missing translation for 'onlineSavingsMsg'
Learn More

Placental Lactogen (hPL) Rabbit Polyclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9067-P1

 View more versions of this product

Product Code. 12643437

  • ONLINE EXCLUSIVE PRICE
    229.00 EUR
    208.83 EUR / 0.50mL
    Save 20.17 EUR
    8% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Human placental lactogen (hPL) is known to originate in the syncytiotrophoblasts. Ultrastructural localization indicates that placental lactogen is stored in small morphologically distinct cytoplasmic granules within these cells and has a different secretory mechanism from other hormonal peptides such as human chorionic gonadotropin. The level of expression of placental lactogen through pregnancy increases gradually to term.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human placental lactogen

Molecular Weight: 21kDa

Positive Control: hPL recombinant protein, Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Placental Lactogen (hPL)
Polyclonal
200µg/mL
Rabbit
500 μL
Steroids Hormones and Growth Factors
Human
IgG
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Unconjugated
Purified human placental lactogen
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.