missing translation for 'onlineSavingsMsg'
Learn More

Pivmecillinam Hydrochloride, LoGiCal

Product Code. 30462130
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462130 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462130 Supplier LoGiCal Supplier No. LGCFOR3230.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pivmecillinam hydrochloride
CAS 32887-03-9
Analyte or Component Names Pivmecillinam hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H33 N3 O5 S . Cl H
Certifications/Compliance ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, hydrochloride (1:1), (2S,5R,6R), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, hydroxymethyl ester pivalate (ester), monohydrochloride, (+)- (8CI), Melysin, Pivmecillinam hydrochloride, Selexid, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, [2S-(2alpha,5alpha,6beta)]-
InChI Formula InChI=1S/C21H33N3O5S.ClH/c1-20(2,3)19(27)29-13-28-18(26)15-21(4,5)30-17-14(16(25)24(15)17)22-12-23-10-8-6-7-9-11-23;/h12,14-15,17H,6-11,13H2,1-5H3;1H/b22-12+;/t14-,15+,17-;/m1./s1
Solution Type Neat
SMILES Cl.CC(C)(C)C(=O)OCOC(=O)[C@@H]1N2[C@H](SC1(C)C)[C@H](\N=C\N3CCCCCC3)C2=O
IUPAC Name 2,2-dimethylpropanoyloxymethyl (2S,5R,6R)-6-[(E)-azepan-1-ylmethylideneamino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 476.03
Quantity 10 mg
Formula Weight 475.1908 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.