Learn More
Eppendorf™ Piston Spring For Research 3 Neo Single Channel Pipettes
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
10 mL
100 μL
2 mL
2.5 to 20 μL
200 μL
300 μL
5 mL
Unit Size:
Each
Description
- Piston springs provide resistance for controlled piston movement, return the piston to its original position after dispensing
- Ensure precision by offering consistent tactile feedback
- Available in various Volume Capacity and color code
Specifications
Specifications
| For Use With | Pipettes |
| Material | Stainless Steel |
| Type | Piston |
| Volume (Metric) | 5 mL |
| For Use With (Equipment) | Pipette Tips |
| Quantity | 1 Pk. |