missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine-d8 Dihydrochloride, TRC

Product Code. 30412568
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412568 10 mg
30419189 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412568 Supplier Toronto Research Chemicals Supplier No. TRCP50850210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H15D8Cl2N5O2
Molecular Weight (g/mol) 432.37
Synonym 5,11-Dihydro-11-[2-(4-methyl-1-piperazinyl-d8)acetyl]-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one Dihydrochloride, Duogastral-d8 Dihydrochloride, Durapirenz-d8 Dihydrochloride, Gasteril-d8 Dihydrochloride, Gastrozepin-d8 Dihydrochloride, Leblon-d8 Dihydrochloride, Maghen-d8 Dihydrochloride, Renzepin-d8 Dihydrochloride, Tabe-d8 Dihydrochloride
SMILES O=C1NC(C=CC=N2)=C2N(C(CN3C([2H])([2H])C([2H])([2H])N(C)C([2H])([2H])C3([2H])[2H])=O)C4=CC=CC=C41.Cl.Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H21N5O2/c1-22-9-11-23(12-10-22)13-17(25)24-16-7-3-2-5-14(16)19(26)21-15-6-4-8-20-18(15)24/h2-8H,9-13H2,1H3,(H,21,26)/i9D2,10D2,11D2,12D2
Chemical Name or Material Pirenzepine-d8 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.