Learn More
Nordic Pack Pipettetes
Product Code. 11381515
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11381515 100 mL Each
11371515 50 mL Each
11361515 30 mL Each
11351515 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11381515 Supplier Nordic Pack Supplier No. 11101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made from polyethylene
  • Available in different capacities
  • Colour: Red
Specifications
Material Polyethylene
Volume (Metric) 100 mL
Type Dropping Pipette