Learn More
Fisherbrand™ Flip & Grip™ Pipette Holder
Product Code. 15380360 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15380360 Each
15291588 Each
15390360 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15380360
  • ONLINE EXCLUSIVE PRICE
    84.60 EUR
    80.20 EUR / Pack of 1
    Save 4.40 EUR
    5% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
Bulk Quote Request
Hang under a shelf to help maximize your working space. Fisherbrand™ Flip & Grip Pipette Holder accommodates 12 single-channel pipettes or a combination of 3 single and 3 multichannel pipettes.
  • Pipettes can be held in the rubber grips and/or the plunger hooks
  • Holds most common brands
  • Rubber grips prevent scratching pipette
  • Holder rotates for easy pipette selection and storage
  • Weighted base
  • Fixing screw provided for under shelf option
Specifications
Autoclavable Not Autoclavable
Holds Single- and Multi-channel Pipettes
Material Metal, Silicone
Shape Circular
Stackable No
Sterility Non-sterile
Product Type Pipette Holder
Shelves None
Height (Metric) 187 mm