Learn More
MP Biomedicals Pipette, 10 to 100 μL
Product Code. 15799160 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
10 to 100 μL
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15799160 10 to 100 μL 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15799160 Supplier MP Biomedicals Supplier No. 07WS1075
  • ONLINE EXCLUSIVE PRICE
    1215.00 EUR
    1157.10 EUR / 1 sheets
    Save 57.90 EUR
    4% Off
Estimated Shipment: 18-12-2026
Positive displacement pipette suited for liquids with high densities, viscosities, or vapor pressures.
  • Problem-free pipetting of problem liquids.
  • Specially suited for liquids with high densities, viscosities or vapor pressures.
  • Given the disposable capillaries and pistons, positive displacement absolutely eliminates cross-contamination from aerosols.
  • Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Specifications
Volume (Metric) 10 to 100 μL
Volume (Metric) Dispensing 10/100 μL
For Use With (Application) Specially suited for liquids with high densities, viscosities, or vapor pressures.
Quantity 1/Pk
Product Type Pipette