Learn More
Gilson™ PIPETMAN™ L Starter Kit
Product Code. 15374765 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15374765 2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15374765 Supplier Gilson™ Supplier No. F167350
  • ONLINE EXCLUSIVE PRICE
    1124.76 EUR
    1097.25 EUR / 1 sets
    Save 27.51 EUR
    2% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
Increases productivity, security and comfort. PIPETMAN™ L starter kit contains three pipettes, conveniently packaged with PIPETMAN DIAMOND Tips and useful accessories.
  • Three PIPETMAN L Pipettes: P20L, P200L, and P1000L with Gilson certificates
  • Two racks of certified-quality PIPETMAN DIAMOND Tips: D200 and D1000
  • Three SINGLE™ pipette holders
  • Three Plastic tip ejectors
  • One Gilson Guide to Pipetting
  • One Two-minute inspection poster
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Blue
Number of Channels 1
Product Line Pipetman L
Type Starter Kit
Volume (Metric) 2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L)