Learn More
Piperidine, Sigma-Aldrich™
Description
Piperidine is a heterocyclic amine with a six-membered ring containing five methylene bridges (-CH2-) and one amine bridge (-NH-). Numerous alkaloids, pharmaceuticals, agrochemicals, and synthetic and biological intermediates contain the piperidine moiety. The commercial applications of piperidine include: solvent for curing agents and epoxy resins. In organic synthesis, it serves as an intermediate.
Specifications
Specifications
| Chemical Name or Material | Piperidine |
| CAS | 110-89-4 |
| Melting Point | -10.8°C |
| Density | 0.86 g/cm³ at 20°C |
| Boiling Point | 106°C (1013 hPa) |
| Flash Point | 16°C (closed cup) |
| Quantity | 170 kg |
| Synonym | Piperidine, Hexahydropyridine, Pentamethyleneimine, Azacyclohexane, Azinane, Piperidine |
| Molecular Weight (g/mol) | 85.15 |
| Grade | Not Available |