Learn More
Piperidine, 99%, extra pure
Product Code. p-7214330 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
100mL
2.5 L
500mL
Product Code. 10159831
Brand: Thermo Scientific Acros undefined
Request Bulk or Custom Format
CAS: 110-89-4 | C5H11N | 85.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
IUPAC Name piperidine
SMILES C1CCNCC1
Specifications
Melting Point -11.0°C
Density 0.8620 g/mL
Boiling Point 106.0°C
Flash Point 16°C
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4515 to 1.4535
Quantity 100 mL
Beilstein 20, 6
Fieser 01,886; 02,332; 04,393; 06,472; 07,293
Merck Index 15, 7580
Specific Gravity 0.862
Solubility Information Solubility aq. soln.: miscible. Other solubilities: soluble in alcohol and benzene, soluble in chloroform, acetone and ethanol
Viscosity 1.46 mPa.s (20°C)
Formula Weight 85.15
Grade Extra Pure
Chemical Name or Material Piperidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P240-Ground/bond container and receiving equipment.