Learn More
Piperazine-2,5-dione, 95%, BLD Pharm™
Product Code. 31033659
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033659 25 g -
31016621 500 g 500g
31020981 100 g 100g
31019215 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31033659 Supplier BLD Pharm Supplier No. BD13219925g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 106-57-0
Molecular Formula C4H6N2O2
MDL Number MFCD00006009
Quantity 25 g
SMILES O=C(CN1)NCC1=O
Molecular Weight (g/mol) 114.10
Percent Purity 95%
Chemical Name or Material Piperazine-2,5-dione