missing translation for 'onlineSavingsMsg'
Learn More

Pipendoxifene hydrochloride, MedChemExpress

Product Code. 16498487
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498487 5 mg 5mg
16480546 20 mg 20mg
16461379 1 mg 1mg
16425828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498487 Supplier Medchem Express Supplier No. HY13724B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) that can be used for the research of breast cancer.

Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) that can be used for the research of breast cancer.

Specifications

Chemical Name or Material Pipendoxifene hydrochloride
CAS 245124-69-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H33ClN2O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (202.82 mM; Need ultrasonic)
SMILES OC1=CC2=C(N(CC3=CC=C(OCCN4CCCCC4)C=C3)C(C5=CC=C(O)C=C5)=C2C)C=C1.Cl
Molecular Weight (g/mol) 493.04
Formula Weight 493.04
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.