Learn More
Brady™ Pipemarkers on a Card
Product Code. 12559737 Shop All Brady Products
Change view
Click to view available options
Display Text:
Air comprimé
Azote
CO2
Eau chaude sanitaire
Eau déionisée
Eau froide sanitaire
Fuel
Hélium
Color:
Black on Yellow
White on Blue
White on Dark Brown
White on Green
Unit Size:
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Color Unit Size
12559737 Air comprimé White on Blue Pack of 5
12589737 CO2 Black on Yellow Pack of 5
12579737 Fuel White on Dark Brown Pack of 5
12599737 Hélium Black on Yellow Pack of 5
12569747 Eau chaude sanitaire White on Green Pack of 5
12529747 Eau déionisée White on Green Pack of 5
12509747 Azote Black on Yellow Pack of 5
12559747 Eau froide sanitaire White on Green Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12559737 Supplier Brady™ Supplier No. N001963
  • ONLINE EXCLUSIVE PRICE
    12.50 EUR
    11.63 EUR / Pack of 5
    Save 0.87 EUR
    6% Off
In Stock
European Standard Pipemarkers reinforce safety and aid maintenance.
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Pipe Markers - Air comprimé
Adhesive Type Permanent Rubber, Permanent, Pressure-sensitive
Color White on Blue
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Air comprimé
For Use With (Application) Pipe and valve marking
Labels per Sheet 5
Languages French
No. per Pack 5 Markers/Card
Quantity 5 Pk.
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0049 in.
Thickness (Metric) 0.124 mm
Width (English) 5.91 in.
Width (Metric) 150 mm
Height (English) 0.47 in.
Height (Metric) 12 mm
Alternate Part Number AIRCOMPRIMÉ150X12CARD-T1-P8