Learn More
Brady™ Pipe Banding Tape
Product Code. 12739864
Change view
Click to view available options
Dimensions (L x W): 33m x 100mm; Color: Dark Brown
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12739864 Dimensions (L x W): 33m x 100mm; Color: Dark Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12739864 Supplier Brady™ Supplier No. 275235
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU
Adhesive Type Acrylic, Permanent
Color Dark Brown
Material Polyester
For Use With (Application) Designed for general identification purposes
Quantity 1 Ea.
Length (Metric) 33 m