Learn More
Ohaus™ Pioneer™ PX 323/E Precision Balance
Product Code. 15855337 Shop All Ohaus Products
Change view
Click to view available options
Capacity: 3200 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15855337 Capacity: 3200 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15855337 Supplier Ohaus™ Supplier No. 30429812
  • ONLINE EXCLUSIVE PRICE
    1175.00 EUR
    874.00 EUR / Pack of 1
    Save 301.00 EUR
    25% Off
View all promotions
In Stock
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding (models with draftshield), and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Not Applicable
Certifications/Compliance CE
Connection Type RS232 & USB
Description Electronic Balance, PX323/E
Capacity 320 g
Display Type Backlit LCD
Readability 0.001000 g
Languages English (US), French, German, Italian, Spanish (MX)
Linearity 0.002000 g
Material ABS Plastic, Cast Metal, Stainless Steel, Glass
Model PX323/E
Repeatability 0.001000 g
Stabilization Time 2.000 sec.
Weighing Modes Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Dimensions (D x W x H) 321 x 209 x 309 mm
Dimensions (L x W x H) 321.00 x 209.00 x 309.000 mm
Length (Metric) 321.00 mm
Width (Metric) 209.00 mm
Height (Metric) 309.000 mm
Depth (Metric) 321.00 mm
Diameter (Metric) Outer Pan 120 mm
Weight (Metric) 4.5 kg
Weight (Metric) Shipping 7 kg
Electrical Requirements 100/240 V ∼200 mA 50/60 Hz 12/18 VA